missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Direct PCR Universal Master Mix

Product Code. 17118430
Change view
Click to view available options
No. of Reactions:
100 Reactions
4 x 500 Reactions
500 Reactions
Unit Size:
34g
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
17118430 100 Reactions 34g
16626332 500 Reactions Each
16616332 4 x 500 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17118430 Supplier Invitrogen™ Supplier No. A44647100
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen Platinum Direct PCR Universal Master Mix is designed to amplify DNA directly from various samples without the need to purify DNA.

Invitrogen Platinum Direct PCR Universal Master Mix is designed to amplify DNA directly from various samples without the need to purify DNA. It contains high-performing, engineered Platinum II Taq Hot-Start DNA Polymerase with dNTPs in an innovative buffer that enables universal primer annealing for superior performance in direct PCR applications.

Features of Platinum Direct PCR Universal Master Mix include:

  • Direct DNA amplification from various samples
  • Universal primer annealing at 60°C
  • Optional Platinum GC Enhancer for specific amplification and improved yields of GC-rich targets
  • Superior specificity, sensitivity, and yields

Platinum Direct PCR Universal Master Mix is designed to work with a variety of samples of different origins such as animal, human, plant, insect, worm, and bacterial samples. The quick lysis protocol enables efficient amplification from templates up to 8 kb in size and co-cycling of different length fragments.

Engineered Platinum II Taq DNA polymerase confers faster cycling and tolerance to reaction inhibitors originating from sample material. Proprietary Platinum Taq antibodies block polymerase activity at ambient temperatures and dissociate after the initial denaturation step. This automatic 'hot start' provides increased sensitivity, specificity, and yield, while allowing reaction assembly at room temperature.

Due to the unique composition of the Platinum Direct PCR Universal Master Mix, the annealing temperature of 60°C can be used for most primer pairs that follow the general design rules. Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair. Different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.

Applications:

  • Direct PCR from animal, plant and bacterial samples
  • Genotyping
  • GMO screening
  • Food and environmental testing
  • CRISPR gene-editing screening
  • Target DNA detection from limited biological material
  • Direct real-time PCR from cell lysates
  • 16S rRNA gene amplification for microbial analysis
  • Colony screening
  • Target amplification from FFPE tissue
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X Platinum Direct PCR Universal Master Mix (1 mL)
• Lysis Buffer (5 mL)
• Proteinase K (150 μL)
• Platinum GC Enhancer (1.25 mL)
• Nuclease-free water (1.25 mL)

Store at –20°C.
GC-Rich PCR Performance High
Polymerase Platinum II Taq Hot-Start DNA Polymerase
Reaction Speed Fast
Product Type Direct PCR Universal Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, Real Time PCR (qPCR)
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Size (Final Product) 8 kb
Show More Show Less
Can I use plant or tissue sample that is not on the list of the Platinum Direct PCR Universal Master Mix tested samples?

You can use any plant or tissue sample. However, when working with new sample materials, we recommend using the Lysis protocol as it allows several PCR reactions to be performed from the same sample during optimization. We also recommend having a positive control with purified sample DNA to ensure that the PCR conditions are optimal. If the positive control with purified DNA fails, the PCR conditions should be optimized before continuing further.

When performing Direct PCR with the Platinum Direct PCR Universal Master Mix, do I need to use Proteinase K?

Proteinase K is required when PCR is performed directly from tissue samples using the Direct PCR protocol. Cell debris present in these PCR products can cause DNA fragments to get trapped in the agarose gel wells and Proteinase K helps to eliminate this.

Is the PCR product from reactions performed with Platinum Direct PCR Universal Master Mix compatible with E-Gel agarose gels?

Electrophoretic separation on E-Gel agarose gels depends on the salt concentration in the analyzed sample. For optimal band separation, we recommend diluting PCR reactions 2- to 20-fold with water, prior to running on E-Gel agarose gels. The dyes in the Platinum Direct PCR Universal Master Mix do not interfere with fragment separation on E-Gel agarose gels.

With the Platinum Direct PCR Universal Master Mix, can I leave the samples in the Lysis Solution (Lysis Buffer containing Proteinase K) for longer than 1 min?

Samples can be incubated for up to 8 hr at room temperature in the Lysis Solution, prior to the 98 degrees C Proteinase K inactivation step. Please note that longer incubation may increase the product yield. For longer incubation, the Proteinase K inactivation step may be increased up to 10 min.

What is the biggest sample size that I can use with Platinum Direct PCR Universal Master Mix?

For the Direct protocol, the recommended sample size is up to 1 mm. For the Lysis protocol, the recommended sample size is up to 2 mm. Sample size can be increased up to 1 cm, however, Lysis Buffer volume needs to be increased to cover the whole sample.

With the Platinum Direct PCR Universal Master Mix, can I adjust the volume of Lysis Buffer depending upon the sample size?

If your sample is larger than 1 mm in diameter, we recommend increasing the Lysis Buffer volume to make sure that the sample is completely covered in the buffer. Add Proteinase K accordingly.

Does the Platinum Direct PCR Universal Master Mix come with a tissue puncher? What kind of tools do I need for sample handling?

No, a tissue puncher is not provided with the kit. If a tissue puncher is not available, cut the sample with a scalpel. Tweezers, scalpel and other tools need to be cleaned with 2% NaClO solution to avoid cross contamination.

What are your recommendations for obtaining a sample from plant or animal tissue for amplification with Platinum Direct PCR Universal Master Mix?

In order to obtain best results when amplifying DNA directly from a sample using Platinum Direct PCR Universal Master Mix, it is important to use a very small amount of starting material. The recommended size is 1-2 mm in diameter. Possible tools for sample handling are: tissue puncher, scalpel or 100 µL pipette tip for soft samples.

Which FFPE sample types can I use with Platinum Direct PCR Universal Master Mix for direct amplification from tissue samples?

FFPE tissue, FFPE tissue sections, FFPE tissue blocks and microscope slides can be used with Platinum Direct PCR Universal Master Mix. DNA is highly fragmented in FFPE samples, and DNA quality being one of the main factors for successful amplification, we recommend amplifying fragments up to 300 bp with this master mix.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.