missing translation for 'onlineSavingsMsg'
Learn More

Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)

Product Code. 11377187
Change view
Click to view available options
Quantity:
25 x 25 mm
Unit Size:
25mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377187 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377187 Supplier Thermo Scientific Alfa Aesar Supplier No. 011507.FF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6-4-7440
Appearance Silvery white foil
Quantity 25 x 25 mm
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Form Foil
Molecular Formula Pt
MDL Number MFCD00011179
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
PubChem CID 23939
ChEBI CHEBI:33400
Chemical Name or Material Platinum
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.