missing translation for 'onlineSavingsMsg'
Learn More

Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis)

Product Code. 11325769
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325769 50 x 50 mm Each
11391358 25 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325769 Supplier Thermo Scientific Alfa Aesar Supplier No. 010282.FI
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 0.47g/25x25mm; Wire cloth; Open area: 62.7%; Width of opening: 0.015 in. | CAS: 7440-06-4 | Pt | 195.08 g/mol

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color White
Appearance Silvery white guage
Quantity 50 x 50 mm
Mesh Size 100 Mesh
Odor Odorless
Assay Percent Range 99.9%
Form Wire Cloth
Solubility Information Insoluble in water. Dissolves in aqua regia
Chemical Name or Material Platinum

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.