missing translation for 'onlineSavingsMsg'
Learn More

Platinum gauze, 45 mesh woven from 0.198mm (0.0078in) dia wire, 99.9% (metals basis)

Product Code. 11314777
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314777 25 x 25 mm Each
11385939 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314777 Supplier Thermo Scientific Alfa Aesar Supplier No. 041814.FF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-06-4 | Pt | 195.08 g/mol

Platinum gauze is used in electronics industry for electrical contacts which might be subjected to high temperatures. It also acts as a catalyst in the production of nitric acid. It plays an essential role in the cracking processes in oil refineries. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 25 x 25 mm
Mesh Size 45 Mesh
Odor Odorless
Weight ≈1.61g/25x25mm
Form Wire Cloth
Solubility Information Soluble in hot aqua regia.
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Platinum gauze, Woven from 0.198mm (0.0078 in.) dia. wire

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.