missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ Tfi Exo(-) DNA Polymerase
Description
Platinum™ Tfi Exo(-) DNA Polymerase is recombinant Tfi Exo(-) DNA Polymerase complexed with a proprietary antibody mix that inhibits polymerase activity at ambient temperature, allowing room-temperature reaction setup. Activity is restored after the initial denaturation step in PCR cycling at 94°C, providing an automatic 'hot start' for increased specificity, sensitivity, and yield. Tfi Exo(-) DNA Polymerase is purified from E. coli. expressing cloned mutants of the Thermus filiformis DNA polymerase gene. This enzyme has 5'→3' polymerase activity, but lacks both 5'→3' and 3'→5' exonuclease activity for improved yield and robustness. Tfi Exo(-) DNA polymerase is heat-stable and will synthesize DNA at elevated temperatures from single-stranded templates in the presence of a primer.
Applications
Platinum™ Tfi Exo(-) DNA Polymerase can be used in protocols that currently use Platinum™ Taq DNA Polymerase without modification. PCR performance is comparable to that of Platinum™ Taq in yield, specificity, fidelity, and robustness. Like Platinum™ Taq, Platinum™ Tfi Exo(-) DNA Polymerase has a nontemplate-dependent terminal transferase activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products.
Applications
Platinum™ Tfi Exo(-) DNA Polymerase can be used in protocols that currently use Platinum™ Taq DNA Polymerase without modification. PCR performance is comparable to that of Platinum™ Taq in yield, specificity, fidelity, and robustness. Like Platinum™ Taq, Platinum™ Tfi Exo(-) DNA Polymerase has a nontemplate-dependent terminal transferase activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products.
Specifications
Specifications
| Concentration | 5X |
| Content And Storage | Platinum™ Tfi DNA Polymerase (500 units total, at 5U/μL) is supplied with 5X Platinum™ Tfi Reaction Buffer (4 × 1.3 mL) and 50 mM Magnesium Chloride (1 mL). Store at -20°C (non-frost-free). |
| GC-Rich PCR Performance | Low |
| Polymerase | Tfi DNA Polymerase |
| Reaction Speed | Standard |
| Product Line | Platinum |
| Product Type | DNA Polymerase |
| Quantity | 500 units |
| Shipping Condition | Dry Ice |
| Fidelity (vs. Taq) | 1 X |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?