missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) acetylacetonate, 98%

Product Code. 11974361 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11974361

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15170-57-7 | C10H14O4Pt | 393.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15170-57-7
Molecular Formula C10H14O4Pt
Molecular Weight (g/mol) 393.30
MDL Number MFCD00000028
InChI Key KLFRPGNCEJNEKU-FDGPNNRMSA-L
Synonym platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
PubChem CID 10960186
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;platinum
SMILES [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 250.0°C to 252.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Linear Formula [CH3COCH=C(CH3)O]2Pt
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, diethyl ether;, soluble in acetone, toluene, diluted acids
Sublimation Point 170 °C
Formula Weight 393.31
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Platinum(II) acetylacetonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Platinum(II) acetylacetonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.