missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ II Hot-Start PCR Master Mixes (2X)

Product Code. 15899844
Change view
Click to view available options
No. of Reactions:
1000 Reactions
20 Reactions
200 Reactions
50 Reactions
Color:
Colorless
Green
Unit Size:
1000 reactions
200 reactions
50 reactions
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Color Unit Size
15899844 200 Reactions Colorless 200 reactions
15929082 20 Reactions Green Each
15815066 50 Reactions Green 50 reactions
15933592 200 Reactions Green Each
15939082 1000 Reactions Green Each
15889844 50 Reactions Colorless 50 reactions
15809854 1000 Reactions Colorless 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15899844 Supplier Invitrogen™ Supplier No. 14000013
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

4x1.25 mL Platinum II PCR Master Mix (2X), 2x1.25 mL Platinum GC Enhancer, 4x1.25 mL Water, nuclease-free

Invitrogen Platinum II Hot-Start PCR Master Mix (2X) offers Platinum II Taq Hot-Start DNA Polymerase premixed with Platinum II PCR buffer and dNTPs for convenient PCR setup.

Invitrogen Platinum II Hot-Start PCR Master Mix (2X), available in colorless and green formats, offers Platinum II Taq Hot-Start DNA Polymerase premixed with Platinum II PCR buffer and dNTPs for convenient PCR setup. Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase and leading hot-start technology. This master mix is provided with the optional Platinum GC Enhancer for specific amplification and improved yields of GC-rich targets.

Platinum II Hot-Start Green PCR Master Mix also includes two tracking dyes for direct loading of PCR products on gels.

Features

  • Innovative buffer enables universal annealing temperature by isostabilizing primer-template duplex structures
  • Engineered Taq DNA polymerase confers fast cycling and resistance to common inhibitors
  • Hot-start technology enables superior specificity, sensitivity, and yields and allows for room temperature reaction setup
  • Green PCR buffer reduces pipetting errors with direct gel loading

Applications:

  • Genotyping and allele screening
  • Amplification from low-input or low-quality DNA
  • GC-rich and AT-rich PCR
  • Sanger sequencing template generation
  • Broad-range microbial/16S amplification
  • Fast and high-throughput PCR
  • Multiplex target amplification
  • Amplification of challenging genomic targets
  • Colony PCR and clone screening
  • qPCR-based pathogen detection and quantification

Platinum II Taq Hot-Start DNA Polymerase is an engineered Taq DNA polymerase that shows increased resistance to reaction inhibitors originating from sample material or DNA purification steps. The polymerase has a higher DNA synthesis rate and delivers PCR results more than two times faster than other Taq DNA polymerases. Proprietary Platinum Taq antibodies block polymerase activity at ambient temperatures and dissociate after the initial denaturation step at 94°C. This automatic hot start provides increased sensitivity, specificity, and yield, while allowing reaction assembly at room temperature.

Due to the unique composition of the Platinum II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer–template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair. With Platinum II Hot-Start PCR Master Mix (2X), different PCR assays can be cycled together using the same protocol with universal primer annealing temperature and the extension step selected for the longest fragment to be amplified.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, colorless format is recommended.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Platinum II PCR Master Mix (2X), 4 x 1.25 mL
• Platinum GC Enhancer, 2 x 1.25 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C in a non-frost-free freezer.
Format Tube
GC-Rich PCR Performance High
Polymerase Platinum II Taq Hot-Start DNA Polymerase
Reaction Speed Fast or Standard
Product Type Hot Start PCR Master Mix
Quantity 200 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Size (Final Product) 5 kb or less
Starting Material DNA
Color Colorless
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.