missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium foil, 0.15mm (0.006in) thick, 99.9% (metals basis)

Product Code. 15426555
Change view
Click to view available options
Quantity:
50 x 50 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426555 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426555 Supplier Thermo Scientific Alfa Aesar Supplier No. 043813.FI
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium foil
Name Note 0.15mm (0.006 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 80:20 wt%
Quantity 50 x 50 mm
Odor Odorless
Weight ≈8.13g/50x50mm
Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD02091683
Percent Purity 99.9%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.