Learn More
Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)
Descrizione
Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Chemical Name or Material | Platinum Iridium foil |
| Name Note | 0.1mm (0.004 in.) thick |
| Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
| Quantity | 50 x 50 mm |
| Odor | Odorless |
| Weight | ≈1.35g/25x25mm |
| Form | Foil |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD02091683 |
| Solubility Information | Insoluble in water. |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.