missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals)

Product Code. 11374658
Change view
Click to view available options
Quantity:
10 cm
50 cm
Unit Size:
10cm
50cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374658 50 cm 50cm
11364658 10 cm 10cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374658 Supplier Thermo Scientific Alfa Aesar Supplier No. 043019.BW
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes.Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Platinum Iridium wire
Name Note 0.127mm (0.005 in.) dia., Annealed, Temper: soft
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 50 cm
Odor Odorless
Weight ≈0.03g/10cm
Form Wire
Assay Percent Notes (metals basis excluding precious metals)
MDL Number MFCD02091683
Solubility Information Insoluble in water and acid.
Percent Purity 99.9%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.