missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)

Product Code. 11388158
Change view
Click to view available options
Quantity:
10 cm
50 cm
Unit Size:
10cm
50cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388158 50 cm 50cm
11378158 10 cm 10cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388158 Supplier Thermo Scientific Alfa Aesar Supplier No. 039383.BW
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Form: approx. 0.11g/10cm,; Temper: soft | CAS: 7440-06-4 | Pt | 195.08 g/mol

Used in the fabrication of metal microelectrodes for electrical stimulation of nervous tissues and in electrophysiological recordings. Inoculating needles and leadwires for implants. Used in electrode materials, composing the gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Density 21.56g/cm3 at 20°C
Quantity 50 cm
Odor Odorless
Assay Percent Range >99.9%
Form Wire
Solubility Information Insoluble in water.
Chemical Name or Material Platinum Iridium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.