missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.5mm (0.02in) dia, hard

Product Code. 11336767
Change view
Click to view available options
Quantity:
10 cm
50 cm
250 cm
Unit Size:
10cm
250cm
50cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336767 10 cm 10cm
11326767 50 cm 50cm
11346767 250 cm 250cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336767 Supplier Thermo Scientific Alfa Aesar Supplier No. 010056.BS
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Form: Approx. 0.44g/10cm, Temper: as drawn | CAS: 7440-6-6 | Pt:Ir; 90:10 wt%

Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
Molecular Formula Pt:Ir; 90:10 wt%
MDL Number MFCD02091683

Specifications

Color White
Appearance Silvery white wire
Quantity 10 cm
Odor Odorless
Form Wire
Solubility Information Insoluble in water
Chemical Name or Material Platinum, Hard

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.