missing translation for 'onlineSavingsMsg'
Learn More

Platinum powder, -200+400 mesh, 99.98% (metals basis)

Product Code. 15447325
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447325 5 g 5g
15457325 100 mg 100mg
15437325 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15447325 Supplier Thermo Scientific Alfa Aesar Supplier No. 046830.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Physical Form Powder
Quantity 5 g
Percent Purity 99.98%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Platinum powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.