missing translation for 'onlineSavingsMsg'
Learn More

Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis)

Product Code. 11376628
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376628 100 x 100 mm Each
11366628 25 x 25 mm Each
11385919 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376628 Supplier Thermo Scientific Alfa Aesar Supplier No. 012217.GH
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platinum-rhodium alloy gauze serves as a catalyst in the oxidation of the ammonia from the Haber Process by atmospheric oxygen to produce nitric oxide and subsequently nitric acid, which is used for producing nitrogen fertilizers, ammonium nitrate, adipic acid, and toluene diisocyanate (precursor for polyurethane). It has high degree of chemical inertness and stability at very high temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Rhodium gauze
Name Note Woven from 0.076mm (0.003 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
Quantity 100 x 100 mm
Mesh Size 80 Mesh
Odor Odorless
Weight ≈0.39g/25x25mm
Assay Percent Notes (metals basis)
MDL Number MFCD00198186
Solubility Information Insoluble in water.
Percent Purity 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.