missing translation for 'onlineSavingsMsg'
Learn More

Platinum Ruthenium foil, 0.05mm (0.002in) thick

Product Code. 11356139
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356139 50 x 50 mm Each
11345427 25 x 25 mm 25mm
11335427 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356139 Supplier Thermo Scientific Alfa Aesar Supplier No. 042686.FI
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pt:Ru; 95.2:4.8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Ruthenium
Color Silver to Gray
Appearance Silver-gary foil
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Molecular Formula Pt:Ru; 95.2:4.8
MDL Number MFCD00798552

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.