Learn More
Platinum standard form crucible, Top OD 32.5mm, Bot Dia 18mm, Ht 36mm, Base Thickness 0.3mm, Capacity 20mL
Product Code. 18867591
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
18867591 1 Ea. Each
1 options
Product Code. 18867591 Supplier Thermo Scientific Alfa Aesar Supplier No. 046520.KT
 Restricted Item
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.