missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SuperFi II PCR Master Mixes

Product Code. 16430725
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
5 x 500 Reactions
500 Reactions
Unit Size:
1 set
100 reactions
2500 reactions
500 reactions
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. of Reactions Unit Size
16430725 Green 5 x 500 Reactions 2500 reactions
16543881 Green 100 Reactions 100 reactions
17155332 Green 500 Reactions 500 reactions
16445389 Colorless 100 Reactions 1 set
16853153 Colorless 500 Reactions 500 reactions
16420725 Colorless 5 x 500 Reactions 2500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16430725 Supplier Invitrogen™ Supplier No. 12369250
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen Platinum SuperFi II PCR Master Mix (2X) is a ready-to-use mixture of Platinum SuperFi II DNA Polymerase, Platinum SuperFi II Buffer, and dNTPs for convenient PCR setup and efficient amplification.

Invitrogen Platinum SuperFi II Green PCR Master Mix (2X) contains a ready-to-use mixture of Platinum SuperFi II DNA Polymerase, Platinum SuperFi II Buffer, and dNTPs for convenient PCR setup, as well as two tracking dyes for direct loading of PCR products on gels. Platinum SuperFi II DNA Polymerase (Cat. No. 12361010) is a proofreading DNA polymerase that combines superior fidelity with innovative SuperFi II Buffer, enabling universal primer annealing for the highest success in PCR. It is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Features of Platinum SuperFi II PCR Master Mix include:

  • Exceptional >300X Taq fidelity
  • Universal primer annealing at 60°C
  • Superior specificity, sensitivity, and yields
  • Robust amplification of difficult-to-amplify targets (e.g., those with suboptimal purity, ˃65% GC content, long PCR requirement)

Platinum SuperFi II DNA Polymerase is an engineered enzyme with high processivity and increased resistance to PCR inhibitors. It also enables fast-cycling protocols and amplification of long targets (up to 20 kb). Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation. This technology also enables reaction set up at room temperature and provides increased sensitivity and yield.

Due to the unique composition of the SuperFi II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize the annealing temperature for each primer pair. With Platinum SuperFi II DNA Polymerase, different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.

Applications of Platinum SuperFi II DNA Polymerase:

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR
  • Fast PCR

Platinum SuperFi II PCR Master Mix (12368010) is also available, which is the same master mix without the tracking dyes.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Five boxes of Platinum SuperFi II Green PCR Master Mix, each containing:
• Platinum SuperFi II Green PCR Master Mix (10 x 1.25 mL)
• Nuclease-free water (10 x 1.25 mL)

Store at –20°C.
GC-Rich PCR Performance High
Polymerase Platinum SuperFi II DNA Polymerase
Reaction Speed Fast
Product Type PCR Master Mix
Quantity 5 x 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) >300X
Hot Start Built-In Hot Start
No. of Reactions 5 x 500 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Color Green
Show More Show Less
How much volume of the Platinum SuperFi II PCR Master Mix (2X) or Platinum SuperFi II PCR Green Master Mix (2X) should I use per reaction?

Typically, for a 50 µL PCR reaction, we recommend using 25µµL of the 2X master mix so that its final concentration in the reaction is 1X.

What is the best way to quantify your PCR samples using the Platinum SuperFi II PCR Master Mix, Green (Cat, No. 12369250, 12369010, 12369050)? Is it possible to quantify using Tapestation or a Qubit fluorometer?

The dyes in the Platinum SuperFi II Green PCR Master Mix can interfere with the fluorescent readings on both Qubit fluorometer and Tapestation. To accurately quantify your samples, you should first use a PCR purification kit, such as the GeneJet PCR Purification Kit (Cat. No. K0701), to clean up your samples. Alternatively, you can use the Platinum SuperFi II PCR Master Mix (Cat. No. 12368010) which does not contain the interfering dyes, thus eliminating the need for a purification step.

What are your recommendations when working with long amplicons using Platinum SuperFi II DNA Polymerase?

Good lab practices are important for long fragment amplification. These include using high-quality templates (pure, fresh, and intact) and fresh primer solutions. Reducing primer concentration to 0.2 µM may also improve the results.

Can I use Platinum SuperFi DNA Polymerase cycling protocol and primer annealing temperature with Platinum SuperFi II DNA Polymerase?

Yes, Platinum SuperFi II DNA Polymerase works at both universal 60 degrees C annealing temperature and at annealing temperatures calculated with a Tm calculator.

Can I perform TA cloning with Platinum SuperFi II DNA Polymerase?

Platinum SuperFi II DNA Polymerase produces blunt-ended PCR products that can be cloned directly into blunt-ended cloning vectors. TA cloning is also possible if 3' dA-overhangs are added after PCR. We recommend purifying the PCR products of Platinum SuperFi DNA Polymerase before adding the overhangs. The procedure for adding 3' dA-overhangs (TA cloning) includes the following steps:
- Purify the PCR product (e.g., with a PCR purification kit or phenol extraction/DNA precipitation). Before adding the overhangs, PCR product must be purified, as the strong proofreading activity of any remaining Platinum SuperFi DNA Polymerase will degrade the added 3' dA-overhangs.
- Perform 3' dA addition with a Taq DNA polymerase.
Reaction components:
Purified PCR product
0.2 mM dATP
1x Taq Buffer
1 U Taq DNA Polymerase
Incubate the reaction for 20 min at 72 degrees C.
- Proceed to TA cloning. For optimal ligation efficiency, we recommend using fresh PCR products, since 3' dA-overhangs can be lost during storage

Can I perform restriction digestion in the Platinum SuperFi II reaction buffer?

Platinum SuperFi II reaction buffer is compatible with restriction digestion directly after PCR. Note that Platinum SuperFi II DNA Polymerase is not inactivated during PCR, thus purification of PCR products before restriction digestion is recommended if intact 5'- or 3'- overhangs are needed for cloning.

Does the green tracking dye in Platinum SuperFi II Green PCR Master Mix interfere with downstream applications?

The green tracking dye (which consists of a blue and a yellow dye) is compatible with downstream applications such as fluorescent automated DNA sequencing, ligation, and restriction digestion. For applications that require analysis of PCR products by absorbance or fluorescence excitation, we recommend using colorless versions of the products (Platinum SuperFi II DNA Polymerase or Platinum SuperFi II PCR Master Mix) or purifying the PCR products prior to analysis.

Will the green tracking dye in Platinum SuperFi II Green PCR Master Mix interfere with fragment analysis in E-Gel agarose gels?

The green tracking dye (which consists of a blue and a yellow dye) does not interfere with electrophoresis in E-Gel agarose gels. The sample should be diluted 2- to 20-fold for optimal separation using E-Gel agarose gels.

Which nucleotide analogues can I use with Platinum SuperFi II DNA Polymerase?

Platinum SuperFi II DNA Polymerase cannot read dUTP-derivatives or dITP in DNA templates, so the use of these analogues is not recommended. Platinum SuperFi II DNA Polymerase can incorporate 7-deaza-dGTP and radiolabeled dNTPs.

Can I amplify AT-rich targets with Platinum SuperFi II DNA Polymerase?

Platinum SuperFi II DNA Polymerase enables amplification of both AT-rich and GC-rich targets. Lower extension temperature (68 degrees C or even 65 degrees C) can be helpful for extremely AT-rich targets.

Can I amplify GC-rich targets with Platinum SuperFi II DNA Polymerase?

Platinum SuperFi II DNA Polymerase can amplify targets with high GC content (up to 75% GC) without any additional DNA melting agents. In cases of extremely GC-rich targets (>75% GC), addition of DMSO to final concentration of 5% is recommended.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.