missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq Green Hot Start DNA Polymerase

Product Code. 15420574
Change view
Click to view available options
Quantity:
600 reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420574 600 reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15420574 Supplier Invitrogen™ Supplier No. 11966034
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen Platinum Taq Green Hot Start DNA Polymerase provides Platinum Taq DNA Polymerase with a 10X Green PCR Buffer.

Invitrogen Platinum Taq Green Hot Start DNA Polymerase provides Platinum Taq DNA Polymerase and a 10X Green PCR Buffer. Platinum Taq DNA Polymerase is a convenient and reliable hot-start thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The hot start property of the enzyme is conferred by thermolabile monoclonal antibodies that render Taq DNA polymerase inactive until the initial PCR denaturation step, thus preventing the extension of nonspecifically annealed primers and improving product yield.

Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary Platinum antibodies. The antibodies dissociate during the initial PCR denaturation step and the DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.

Platinum Taq Green Hot Start DNA Polymerase is provided with Green PCR Buffer that contains a density reagent and two tracking dyes and allows for direct gel loading of PCR products. The optional KB Extender used with Platinum Taq DNA Polymerase enables more versatility in PCR assays with long or GC-rich amplicons. Use Platinum Taq Green Hot Start DNA Polymerase for the amplification of DNA from complex genomic, viral, and plasmid templates, as well as in RT-PCR.

Features

  • High specificity and increased yields with antibody-mediated hot-start PCR
  • Convenient room-temperature reaction setup
  • Direct gel loading with green PCR buffer
  • Versatile formulation for broad range of amplicons

Notes

  • The next-generation enzyme Platinum II Taq Hot-start DNA Polymerase is recommended for superior performance.
  • Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Platinum Taq DNA Polymerase, 120 μL
• 10X Green PCR Buffer (without magnesium), 3 x 1.25 mL
• 50 mM MgCl2, 1.25 mL
• KB Extender, 1.25 mL

Store at -20°C in a non-frost-free freezer.
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Quantity 600 reactions
Shipping Condition Dry Ice
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 600 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Show More Show Less
Do you have any recommendations for troubleshooting the amplification of GC-rich or problematic targets with Platinum Taq Green Hot Start DNA Polymerase?

We have the following recommendations:
- Increase the extension time from 1 min/kb to 1.5 min/kb.
- Vary total PCR cycles from 20-40; 35 cycles is typical.
- Try denaturing at 95 degrees C for 45 seconds.
- Increase the amount of template for targets >5 kb.
- Use 2.5 U of Platinum Taq for each 50 µL reaction.
- GC-rich or problematic targets work better with MgSO4 instead of MgCl2.
- For GC-rich templates, test higher annealing temperatures starting with the temperature that is equal to your primer Tm and in increments of 2 degrees C for up to six different temperatures.
- Vary the KB extender solution from 1.5 µL to 4.5 µL per 50 µL reaction.

When should the KB Extender be used that comes with the Platinum Taq Green Hot Start DNA Polymerase (Cat. Nos. 11966018, 11966026, 11966034, 11966083)?

The KB Extender enhances the amplification of GC-rich and problematic sequences, and the extension of genomic DNA targets of >5 kb. The KB Extender lowers DNA melting temperature (Tm), and the co-solvent and amplification buffer offer higher primer specificity, broader Mg concentration and annealing temperature optima, as well as improved Taq thermostabilization. Please note that primer sets that generate specific products using standard PCR buffer may not benefit from using KB Extender. Excessive use of the KB Extender may reduce yield, particularly for non-GC rich amplicons. We generally recommend varying the KB Extender solution from 1.5-4.5 µL per 50 µL reaction.

Is the green dye that comes with the Platinum Taq Hot Start Green DNA Polymerase and Platinum Green Hot Start PCR Master Mix (2X) compatible with downstream applications?

Yes the green dye is compatible with downstream applications such as fluorescent automatic DNA sequencing, ligation, and restriction digestion.

Do you offer Platinum Taq DNA Polymerase formats that already contain loading dyes in the reaction buffer?

Yes, we offer several Platinum Taq DNA polymerases and their master mixes, supplemented with tracking dyes for direct loading of PCR products on gels:

- Platinum II Hot-Start Green PCR Master Mix (2X)
- Platinum SuperFi Green DNA Polymerase and Platinum SuperFi Green PCR Master Mix
- Platinum Taq Green Hot-Start DNA Polymerase and Platinum Green Hot-Start PCR Master Mix (2X)
- DreamTaq Green Hot-Start DNA Polymerase and DreamTaq Hot-Start Green PCR Master Mix
- Phusion Green Hot-Start II High-Fidelity DNA Polymerase and Phusion Green Hot-Start II High-Fidelity PCR Master Mix.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.