missing translation for 'onlineSavingsMsg'
Learn More

Platinum Wire, 0.076mm (0.003 in.) dia., Thermo Scientific™

Product Code. 11356777
Change view
Click to view available options
Quantity:
50 cm
2 m
Unit Size:
2m
50cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356777 2 m 2m
11346777 50 cm 50cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356777 Supplier Thermo Scientific Alfa Aesar Supplier No. 010079.G2
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Form: Approx .0.045g/50cm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
Molecular Formula Pt:Rh; 70:30 wt%
MDL Number MFCD00198186

Specifications

Color Silver
Appearance Silver colored wire
Quantity 2 m
Odor Odorless
Form Wire
Chemical Name or Material Platinum, 30 wt% Rhodium wire
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.