Learn More
Olympus™ PLCN 100XO 00X Plan C Achromat Oil-Immersion Objective
Product Code. 15431495
Change view
Click to view available options
Magnification Power:
100X
Unit Size:
Each
Product Code. Magnification Power unitSize
15431495 100X Each
1 options
Product Code. 15431495 Supplier Olympus™ Supplier No. N4239400
Corrected for flat field, spherical, and chromic aberration.
  • Corrected for axial chromatic aberration.
  • Corrected for spherical aberration.
  • Corrected for flatness of field.
Specifications
Type Oil-immersion, Plan C Achromat
Connector Types RMS Threading
Description Field number: 22
Focus Distance Working distance: 0.13 mm
Magnification Power 100X
Numerical Aperture 1.25