missing translation for 'onlineSavingsMsg'
Learn More

Mutexil™ Non-woven Protective Clip Caps

Product Code. 11760114
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11760114 Green Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11760114 Supplier Mutexil™ Supplier No. 4561334
  • ONLINE EXCLUSIVE PRICE
    76.65 €
    27.02 € / Pack of 1000
    Save 49.63 €
    64% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Non-woven clip caps provide basic protection. The elastic headband is encapsulated in soft poly thread for worker comfort.

  • Polypropylene
  • Disposable
  • Bouffant style
  • Pleated

Specifications

Product Type Peaked Bouffant Cap
Size 24 in.
Color Green
Disposable Single-use
Quantity 1000/Pk.
Diameter (Metric) 61 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.