missing translation for 'onlineSavingsMsg'
Learn More

Pleconaril, TRC

Product Code. 30136080
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136080 10 mg 10mg
18021430 50 mg 50mg
18042630 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136080 Supplier Toronto Research Chemicals Supplier No. TRCP58035010MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153168-05-9
Molecular Formula C18 H18 F3 N3 O3
Molecular Weight (g/mol) 381.35
Synonym 3-[3,5-Dimethyl-4-[3-(3-methyl-5-isoxazolyl)propoxy]phenyl]-5-(trifluoromethyl)-1,2,4-oxadiazole, Picovir, Pleconaril, VP 63843, Win 63843
IUPAC Name 3-[3,5-dimethyl-4-[3-(3-methyl-1,2-oxazol-5-yl)propoxy]phenyl]-5-(trifluoromethyl)-1,2,4-oxadiazole
SMILES Cc1cc(CCCOc2c(C)cc(cc2C)c3noc(n3)C(F)(F)F)on1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H18F3N3O3/c1-10-7-13(16-22-17(27-24-16)18(19,20)21)8-11(2)15(10)25-6-4-5-14-9-12(3)23-26-14/h7-9H,4-6H2,1-3H3
Formula Weight 381.13
Chemical Name or Material Pleconaril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.