missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti6.3/V5-DEST™ Gateway™ Vector Kit

Product Code. 10647015
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10647015 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10647015 Supplier Invitrogen™ Supplier No. V53306
  • ONLINE EXCLUSIVE PRICE
    4095.00 €
    3357.90 € / Pack of 1
    Save 737.10 €
    18% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pLenti6.3/V5-DEST™ Gateway™ Vector Kit

Invitrogen™ 's pLenti6.3⁄V5-DEST™ Gateway™ Vector Kit is part of our ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit (catalog K5330-00).

The pLenti6.3⁄V5-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ HiPerform™ lentiviral expression vector, pLenti6.3⁄V5-DEST™ for easy recombination-based cloning and high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti6.3⁄V5-DEST™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements.

Advantages
  • Stable expression
  • Long-term experiments
  • Accurate titer of functional virus
  • Flexible and versatile Gateway™ recombination cloning technology

    Key Features
  • HiPerform™ WPRE and cPPT elements
  • CMV promoter
  • V5 epitope tag at C terminus
  • Blasticidin selection

    Kit includes
  • A lacZ vector as a positive control, pLenti6.3⁄V5-GW⁄lacZ
  • Stbl3™ competent cells

    For research use only. Not intended for any therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Constitutive or Inducible System Constitutive
    Delivery Type Lentiviral
    Promoter CMV
    Product Type Kit, Vector
    Selection Agent (Eukaryotic) Blasticidin
    Content And Storage Vector Kit (Box 1)
    pLenti6.3⁄V5-DEST™ (150 ng⁄μL): 6 μg, store at -20°C
    pLenti6.3⁄V5-GW⁄lacZ Control vector (500 ng⁄μL): 10 μg, store at -20°C

    One Shot™ Stbl3™ Chemically Competent E. coli (Box 2)
    Stbl3™ Cells: 21 x 50 μL, store at -80°C
    pUC19 control DNA (10 pg⁄μL): 50 μL, store at -80°C
    S.O.C. Medium: 6 mL, store at -80°C
    Cloning Method Gateway
    Quantity 1 kit
    Vector pDEST, pLenti
    Product Line Gateway, HiPerform, ViraPower
    For Use With (Application) Viral Expression
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.