Learn More
Memmert™ Plug-in Tube Extensions for Oven
Product Code. 15165586 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
Exhaust Air Ducting
Models U/UF TS/UNpa/S/I/ICP/IPP/IPS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15165586 Exhaust Air Ducting Each
15125543 Models U/UF TS/UNpa/S/I/ICP/IPP/IPS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15165586 Supplier Memmert™ Supplier No. B29719
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use these tube extensions with Memmert ovens. Memmert™ Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.
  • Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
  • Angled
  • Only models U, I, S
Specifications
For Use With (Equipment) Exhaust Air Ducting
Type Plug-in Tube Extension
Diameter (English) 2.2 (Inside) in., 2.4 (Outside) in.
Diameter (Metric) 57 (Inside) mm, 60.3 (Outside) mm