missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments

Product Code. 12886036 Shop All Thermo Scientific Products
Change view
Click to view available options
Needle Gauge:
23 G
26 G
31 G
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
12886036 23 G Each
10310975 31 G Each
10585245 26 G Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12886036 Supplier Thermo Scientific™ Supplier No. 365A0241
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
  • TRUSTED_SUSTAINABILITY

    Specifications

    Description 0.5 μL volume; 23 gauge; 57 mm length
    Needle Gauge 23 G
    Length (Metric) 57 mm
    Product Type GC Autosampler Syringe, Removable Needle
    Disposable Reusable
    Quantity Ea.
    Volume (Metric) 0.5 μL
    Length (Metric) Needle 57 mm
    Needle Point Style Cone
    For Use With (Equipment) TriPlus RSH Autosampler and TriPlus 100LS Autosampler
    Connection Type Removable
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.