missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pluronic F-68 Polyol, 10% Solution (100X)

Product Code. 11495005 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495005 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495005 Supplier MP Biomedicals™ Supplier No. 092750049
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Non-ionic surfactant in cell culture used to control shear forces in suspension cultures against possible damage.

Pluronic F-68 is a difunctional block copolymer surfactant terminating in primary hydroxyl groups. It is a non-ionic detergent that protects cells from hydrodynamic damage, including freezing and thawing, transfer and stirring. It can also be used to reduce foaming in stirred cultures and reduce cell attachment to glass. Used for in vitro isolation of lymphocytes from diluted whole blood. Pluronic F-68™ is a registered trademark of BASF Corporation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pluronic F-68™ Polyol
Quantity 100 mL
Synonym 10% Solution, 100X

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.