missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PlusOne DNA Silver Staining Kit

Product Code. 10208714 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
4mL
Product Code. Quantity unitSize
10208714 1 Kit 4mL
1 options
This item is not returnable. View return policy

Product Code. 10208714

Brand: Cytiva 17600030

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Minimizes production of toxic waste and optimizes sensitivity and robustness making the kit especially suitable for use in automatic staining equipment

  • Typical sensitivity: 20 to 50pg DNA/band
  • Reaction time: <1.5 hr.
  • Kit contents: fixing solution, staining solution, sodium carbonate solution, 37% formaldehyde, 2% sodium thiosulfate, stopping and preserving solution, instruction booklet, short instructions (laminated sheet)

TRUSTED_SUSTAINABILITY

Specifications

Description DNA Silver Staining Kit provides reproducible and sensitive nonradioactive detection of nucleic acids in polyacrylamide gels in less than 1.5 h. Brings speed, convenience and very high sensitivity to the staining of proteins and nucleic acids.
Form Liquid
For Use With (Application) Automatic staining equipment, Standard manual Silver staining of DNA in Polyacrylamide gels
Kit Contents Fixing solution 5X: Benzene Sulphonic Acid, 3% w/v in 24% v/v Ethanol 500mL, Staining solution 5X: Silver Nitrate, 1% w/v Benzene Sulphonic Acid, 0.35 % w/v 500mL, Sodium Carbonate solution 5X: Sodium Carbonate, 12.5% w/v 500mL, Formaldehyde 37%
Quantity 1 Kit
Storage Requirements 10° to 30°C
Product Type DNA Silver Staining Kit
Stain Type Silver Stain
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1
  • Respiratory sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.