Learn More
ERLAB PMMA Cable Ducts
Product Code. 30484795 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Exhaust and Auxiliary Air Duct
Hoods and Enclosures
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
30484795 Hoods and Enclosures Each
30484792 Exhaust and Auxiliary Air Duct Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30484795 Supplier ERLAB Supplier No. NORT000476/D
  • ONLINE EXCLUSIVE PRICE
    196.00 €
    180.00 € / Pack of 1
    Save 16.00 €
    8% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Organize and protect cables with PMMA Cable Ducts
  • Cable ducts allow clean, safe installation of electrical cables in the hood workspace
  • Provide chemical resistance and visual inspection of cables
  • Avoid cable clutter and protect against mechanical wear and contamination
  • Mounted on the right side of the hood for ergonomic access and optimal cable management
  • Improve airflow fluidity by reducing obstruction caused by cables entering directly through the chassis
  • Ideal for configurations involving multiple instruments, sensors or monitoring devices
Specifications
For Use With (Equipment) Hoods and Enclosures
Product Type Upper Duct Transition Adapter