Learn More
ERLAB PMMA Cable Ducts
Product Code. 30484792
Click to view available options
For Use With (Equipment):
Exhaust and Auxiliary Air Duct
Hoods and Enclosures
Unit Size:
Each
Product Code. 30484792
Brand: ERLAB NORT000476/G
Organize and protect cables with PMMA Cable Ducts
  • Cable ducts allow clean, safe installation of electrical cables in the hood workspace
  • Provide chemical resistance and visual inspection of cables
  • Avoid cable clutter and protect against mechanical wear and contamination
  • Mounted on the right side of the hood for ergonomic access and optimal cable management
  • Improve airflow fluidity by reducing obstruction caused by cables entering directly through the chassis
  • Ideal for configurations involving multiple instruments, sensors or monitoring devices
Specifications
For Use With (Equipment) Exhaust and Auxiliary Air Duct
Product Type Upper Duct Transition Adapter