missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PMP Class A Measuring Flask
PMP flask with polypropylene stopper
Brand: BRAND™ 36180
Code : NEW
Additional Details : Weight : 0.07000kg
Description
Specifications
Autoclavable | |
PMP | |
Ground joint: NS 19/26 |
500 mL | |
PP stopper | |
0.25 mL |