missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PMSF

Product Code. 18752754
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752754 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 18752754 Supplier R&D Systems™ Supplier No. 4486/50
  • ONLINE EXCLUSIVE PRICE
    105.38 €
    93.45 € / 50mg
    Save 11.93 €
    11% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

A Tocris product, part of R&D Systems. Non-selective protease inhibitor; also blocks anandamide metabolism

Non-selective protease inhibitor; irreversibly sulfonates key active site serine residues in serine proteases, acetylcholinesterases, and thiol proteases. Blocks metabolism of anandamide (Cat. No. 1339) in a rodent model of cannabimimetic activity. Unstable in water (t1/2 ∽ 60 mins at pH 7.5). Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 329-98-6
Quantity 50 mg
Product Line Tocris
Molecular Formula C7H7FO2S
Formula Weight 174.19
Purity >98%
Molecular Weight (g/mol) 174.189
Chemical Name or Material PMSF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.