missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress PNPP disodium

Product Code. 16481579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481579 1 g 1g
16469457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481579 Supplier MedChemExpress Supplier No. HY15928
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PNPP (Para-nitrophenyl phosphate) disodium is a non-proteinaceous chromogenic substrate for alkaline and acid phosphatases used in ELISA and conventional spectrophotometric assays.

Specifications

Quantity 1 g
Storage Requirements 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Solubility H2O : 160 mg/mL (608.25 mM; Need ultrasonic)
Shipping Condition Room temperature
CAS 4264-83-9
Molecular Weight (g/mol) 263.05 g/mol
Synonym Para-nitrophenyl phosphate disodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.