missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Product Code. 11922281
Click to view available options
Volume (Metric):
100 nL
250 nL
50 nL
Sterility:
Non-sterile
Sterile
Packaging:
20 x 384 tips
20 x 96 tips
missing translation for 'unitSize'
Pack of 3840
Pack of 960
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11922281

missing translation for 'mfr': Thermo Scientific™ 5757100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing.

Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.

Features:

  • Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
  • Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
  • Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
  • Control consistent accuracy and precision during the molding process
Validation and Compatibility:
  • Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
  • With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
  • Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
  • Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
Ordering Information:
  • Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
  • Sterile format available for sensitive liquid handling procedures
TRUSTED_SUSTAINABILITY

Spezifikation

Volume (Metric) 100 nL
Filtered Non-filtered
Sterility Sterile
Format Rack
Tip Style Automation
Packaging 20 x 384 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus
Product Line PocketTip

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt