missing translation for 'onlineSavingsMsg'
Learn More

Podophyllotoxin, TRC

Product Code. 30409748
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409748 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409748 Supplier Toronto Research Chemicals Supplier No. TRCP68100050MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Podophyllotoxin
CAS 518-28-5
Molecular Formula C22 H22 O8
Quantity 50 mg
Synonym (5R,5aR,8aR,9R)-5,8,8a,9-tetrahydro-9-hydroxy-5-(3,4,5-trimethoxyphenyl)-Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one','5,8,8a,9-tetrahydro-9-hydroxy-5-(3,4,5-trimethoxyphenyl)-','[5R-(5α,5aβ,8aα,9α)]-, Podophyllotoxin (6CI,8CI), (-)-Podophyllotoxin, Condyline, Condylox, NSC 24818, Podofilox, (5R,5aR,8aR,9R)-9-Hydroxy-5-(3,4,5-trimethoxyphenyl)-5,8,8a,9-tetrahydro[2]benzofuro[5,6-f][1,3]benzodioxol-6(5aH)-one
InChI Formula InChI=1S/C22H22O8/c1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h4-7,13,18-20,23H,8-9H2,1-3H3/t13-,18+,19-,20-/m0/s1
SMILES COc1cc(cc(OC)c1OC)[C@H]2[C@@H]3[C@H](COC3=O)[C@@H](O)c4cc5OCOc5cc24
IUPAC Name (5R,5aR,8aR,9R)-5-hydroxy-9-(3,4,5-trimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
Molecular Weight (g/mol) 414.41
Formula Weight 414.1315
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.