Learn More
Motic™ Polarizing Set, N2GG Stereo Microscope Stand
Product Code. 11732646
Change view
Click to view available options
For Use With (Equipment):
N2GG Stand
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11732646 N2GG Stand Each
1 options
Product Code. 11732646 Supplier Motic™ Supplier No. 1101000600651
  • ONLINE EXCLUSIVE PRICE
    109.00 €
    93.84 € / Pack of 1
    Save 15.16 €
    13% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Improve image contrast with a polarization set containing a polarizer and analyzer. Motic™ Polarizing Set, N2GG Stereo Microscope Stand permits 360° of rotation to accommodate observation demands.
  • Improve image quality obtained with birefringent materials when compared to other techniques such as darkfield and brightfield illumination, differential interference contrast, phase contrast, Hoffman modulation contrast, and fluorescence.
  • Useful for both quantitative and qualitative studies targeted at a wide range of anisotropic specimens.
  • Polarized light microscopy is capable of providing information on absorption color and optical path boundaries between minerals of differing refractive indices, in a manner similar to brightfield illumination, but the technique can also distinguish between isotropic and anisotropic substances.
  • Contrast-enhancing technique exploits the optical properties specific to anisotropy and reveals detailed information concerning the structure and composition of materials that are invaluable for identification and diagnostic purposes.
Specifications
Type Polarizer, Analyzer
Description Permits 360° of rotation to accommodate observation demands.
For Use With (Equipment) N2GG Stand