missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PolarScreen™ Androgen Receptor Competitor Assay Kit, Green
Description
The PolarScreen™ Androgen Receptor Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential androgen receptor (AR) ligands. The kit uses insect cell-expressed, rat AR-ligand binding domain tagged with glutathione-S-transferase (GST) and histidine (also available separately), plus a novel, high-affinity, fluorescent androgen ligand (Fluormone™ AL Green) in a homogenous, mix-and-read assay format.
This kit produces a green color read-out; a 'red' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.
The PolarScreen Androgen Receptor Competitor Assay is:
Complete & ready-to-use out of the box—all reagents included, just add your test compounds
High throughput compatible—fluorescence polarization assays in a 384-well plate
Convenient—components available in bulk sizes for larger screening needs
How the Assay Works
The kit uses the rat AR ligand-binding domain tagged with His and GST [AR-LBD(His-GST)]. AR-LBD(His-GST) is added to a fluorescently-tagged androgen ligand (FluormoneAL Green) in the presence of competitor test compounds in microwell plates. The presence of effective competitors prevents the formation of a AL Green/AR-LBD(His-GST) complex, resulting in a decrease of the polarization value due to ligand displacement caused by a competitor. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for AR-LBD(His-GST).
Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
This kit produces a green color read-out; a 'red' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.
The PolarScreen Androgen Receptor Competitor Assay is:
How the Assay Works
The kit uses the rat AR ligand-binding domain tagged with His and GST [AR-LBD(His-GST)]. AR-LBD(His-GST) is added to a fluorescently-tagged androgen ligand (FluormoneAL Green) in the presence of competitor test compounds in microwell plates. The presence of effective competitors prevents the formation of a AL Green/AR-LBD(His-GST) complex, resulting in a decrease of the polarization value due to ligand displacement caused by a competitor. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for AR-LBD(His-GST).
Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
Specifications
Specifications
| Quantity | 800 x 20 μL assays |
| Content And Storage | 1 tube AR Ligand Binding Domain GST/His-tagged Recombinant Protein (store at -68 to -85°C) 1 tube of Fluormone AL Green (store at -5 to -30°C) 2 bottles of Screening Buffer (store at -5 to -30°C) 1 tube of 1 M DTT (store at -68 to -85°C) |
| Product Line | PolarScreen |
| For Use With (Application) | Competitive Binding Assay, Nuclear Receptor Assay |
| Ligand | AR |
| Product Type | Assay Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?