missing translation for 'onlineSavingsMsg'
Learn More

Polidocanol, MedChemExpress

Product Code. p-200099712
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16462369

Brand: Medchem Express HYB2106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Polidocanol is a sclerosing agent used successfully to treat extremity and esophageal varices and telangiectasias.

Polidocanol is a sclerosing agent used successfully to treat extremity and esophageal varices and telangiectasias.

Specifications

Chemical Name or Material Polidocanol
CAS 9002-92-0
Color White
Purity Grade Notes Research
Molecular Formula (C2H4O)nC12H26O
Quantity 1 g
Synonym Polyoxyethylene lauryl ether Polyoxyethyleneglycol Dodecyl Ether
Solubility Information H2O : 50 mg/mL (Need ultrasonic) ∣DMSO : 50 mg/mL (Need ultrasonic)
SMILES OCCCOCCCCCCCCCCCCC.[n]
Percent Purity 97.0%
Grade Research
Physical Form Crystals
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.