Learn More
Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar 044745.02
Quantity 0.5g
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15426615
  • 516.00 € / 0.5g
 Restricted Item
In Stock
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Identifiers
104934-50-1
196.352
DUFPJSOXRHVDOV-UHFFFAOYSA-N
22950249
CCCCCCC1=C(SC(=C1)C)C
C12H20S
MFCD00217686
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
3-hexyl-2,5-dimethylthiophene
Specifications
104934-50-1
MFCD00217686
Soluble in Chloroform and tetrahydrofuran
DUFPJSOXRHVDOV-UHFFFAOYSA-N
3-hexyl-2,5-dimethylthiophene
22950249
C12H20S
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
0.5g
CCCCCCC1=C(SC(=C1)C)C
196.352
Poly (3-hexylthiophene-2,5-diyl), Regioregular, low metals
Product Suggestions
Product Suggestions
Videos
SDS Documents