missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-octylthiophene-2,5-diyl), regiorandom, low metals, Thermo Scientific Chemicals

Product Code. 11460041
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460041 1 g 1g
11450041 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460041 Supplier Thermo Scientific Alfa Aesar Supplier No. 044778.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Conducting polymers

Conducting polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104934-51-2
Molecular Formula (C12H18S)n
Molecular Weight (g/mol) 241.44
MDL Number MFCD00241412
InChI Key RXIQSINDOTYIGV-UHFFFAOYSA-N
Synonym poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
PubChem CID 121233643
IUPAC Name azane;2,5-dimethyl-3-octylthiophene
SMILES CCCCCCCCC1=C(-*)SC(-*)=C1

Specifications

Physical Form Powder
Solubility Information Soluble in THF, CHCl3
Quantity 1 g
Chemical Name or Material Poly (3-octylthiophene-2,5-diyl), Regiorandom, low metals

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.