missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Poly(A) Polymerase, Yeast

Product Code. 15960123
Change view
Klicka för att se tillgängliga alternativ
Quantity:
25 KU
Förpackningsstorlek:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
15960123 25 KU Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15960123 Leverantör Thermo Scientific™ Leverantörsnummer 74225Z25KU

Please to purchase this item. Need a web account? Register with us today!

Edge
Denna artikel kan inte returneras. Se returpolicy

Catalyses the addition of adenosine residues onto the 3' ends of RNA. It can be used to add poly(A) tails to RNA in the first step of cloning.

Poly(A) Polymerase catalyses the addition of adenosine residues onto the 3' ends of RNA. It can be used to add poly(A) tails to RNA in the first step of cloning. The reaction requires Mn2+ or Mg2+, ATP as substrate, and any RNA containing 3' hydroxyl termini as primer. Substitution of cordycepin-5'-triphosphate (3'-dATP) for ATP results in addition of a single 3'-dA residue to the ends of the RNA, a useful technique for labeling RNA at the 3' end.

In comparative studies, yeast poly(A) polymerase works more efficiently than E. coli poly(A) polymerase for RNA oligonucleotide-labeling and poly(A) tailing. Shorter incubation times are required for the yeast enzyme and it is found to label both long and short substrates equally well. Poly(A) Polymerase is recommended over T4 RNA Ligase for 3'-end labeling of long RNA molecules.

*Note: Various modified nucleotides can also be used to label the 3' end of RNA using Yeast Poly(A) Polymerase.

Purity:

Ribonuclease free

Storage Buffer:

20 mM Tris-HCl (pH 8.0), 50 mM KCl, 0.5 mM DTT, 50% glycerol.

Assay Conditions:

2 5mM Tris-HCl (pH 7.0), 40 mM KCl, 0.5 mM MnCl2, 0.05 mM EDTA, 0.5 mM DTT, 0.2 mg/mL BSA, 10% glycerol, 3.3 μM radiolabeled ATP, 0.5mM ATP, 6.5 μg poly(A) (∼100 bases), poly(A) polymerase. After incubation at 37°C for 10 min, acid insoluble radioactivity is determined.

Unit Definition:

One unit equals 15 pmol/min AMP incorporated into (riboA)15 at 37°C.

Concentration:

600 units/μL

Functional Test:

3' -end labeling of a ribonucleotide with cordycepin-5'-triphosphate.

Functionally Tested 5X Poly(A) Polymerase Reaction Buffer (1 mL included, PN 74226):

100 mM Tris-HCl, pH 7.0, 3.0 mM MnCl2, 0.1 mM EDTA, 1mM DTT, 500 μg/mL acetylated BSA, 50% glycerol.

Applications:

  1. Addition of poly(A) tails to RNA.
  2. Labeling the 3' ends of RNA.

Source:

E. coli strain containing an overproducing clone of Yeast Poly(A) Polymerase.

TRUSTED_SUSTAINABILITY

Specifikationer

Content And Storage Shipped on dry ice. Store at -20°C.
Polymerase Poly(A) RNA Polymerase
Quantity 25 KU

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.