missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals

Product Code. 12347113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12347113 250 g Glass bottle 250g
12337113 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12347113 Supplier Thermo Scientific Acros Supplier No. 448532500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62649-23-4
Molecular Formula (C3H5NO)n
Molecular Weight (g/mol) 237.19
MDL Number MFCD00084393
InChI Key DTBMFUOWAASNDB-UHFFFAOYSA-M
Synonym 2-propenamide, polymer with 2-propenoic acid and sodium 2-propenoate, sodium acrylamide acrylic acid acrylate
PubChem CID 23696299
IUPAC Name sodium;prop-2-enamide;prop-2-enoate;prop-2-enoic acid
SMILES NC(=O)C(-*)C-*

Specifications

Packaging Glass bottle
Quantity 250 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, formamide, glycerol
Chemical Name or Material Poly(acrylamide), carboxyl modified

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.