missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylamide), carboxyl modified, pure, low carboxyl content, ∽ MW 200,000, Thermo Scientific™

Product Code. 10771841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771841 250 g Glass bottle 250g
10686572 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10771841 Supplier Thermo Scientific Acros Supplier No. 184232500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 62649-23-4
Molecular Formula (C3H5NO)n
Molecular Weight (g/mol) 237.19
MDL Number MFCD00084393
InChI Key DTBMFUOWAASNDB-UHFFFAOYSA-M
Synonym 2-propenamide, polymer with 2-propenoic acid and sodium 2-propenoate, sodium acrylamide acrylic acid acrylate
PubChem CID 23696299
IUPAC Name sodium prop-2-enamide prop-2-enoic acid prop-2-enoate
SMILES NC(=O)C(-*)C-*

Specifications

Color White
Physical Form Powder or Granules
Quantity 250 g
Formula Weight ≈200.000
Grade Pure
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Chemical Name or Material Poly(acrylamide), carboxyl modified, Carboxyl Modified, Low Carboxyl Conten

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.