missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), 25 wt% soln. in water

Product Code. 11419870
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419870 100 g 100g
11429870 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419870 Supplier Thermo Scientific Alfa Aesar Supplier No. 044669.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol

Polyacrylic acid is used in disposable diapers and in ion exchange resins. It is also used to study solute diffusion in polyvinyl alcohol/polyacrylic acid copolymer hydrogel. It is also employed as a thickening, suspending, emulsifying and dispersing agent in pharmaceuticals, cosmetics, adhesives and paints. Further, it is used for the preparation of poly(N-isopropylacrylamide)-block-polyacrylic acid copolymer which responds to both temperature and pH stimuli. In addition to this, it is used in preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-01-4
Molecular Formula (C3H4O2)n
Molecular Weight (g/mol) 72.06
MDL Number MFCD00084394
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name Poly(acrylic acid)
SMILES OC(=O)C(-*)C-*

Specifications

Physical Form Liquid
Density 1.09 g/mL
Quantity 100 g
Solubility Information Soluble in water.
Concentration or Composition (by Analyte or Components) 25 wt% soln. in water
Chemical Name or Material Poly (acrylic acid)
What is the average molecular weight of Poly(acrylic acid), 25 wt% soln. in water?

The average molecular weight of the Poly(acrylic acid), 25 wt% soln. in water is 240,000 g/mole.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.