missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000

Product Code. 10133820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133820 250 g Glass bottle 250g
10337460 5 g Glass bottle 5g
10082380 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133820 Supplier Thermo Scientific Acros Supplier No. 184992500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-01-4 , 79-10-7 , 7732-18-5
Molecular Formula (C3H4O2)n
Molecular Weight (g/mol) 72.06
MDL Number MFCD00084394
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
SMILES OC(=O)C(-*)C-*

Specifications

Physical Form Liquid
Boiling Point 100.0°C
Linear Formula [-CH2CH(CO2H)-]n
Density 1.2300g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane
Quantity 250 g
IUPAC Name Poly(acrylic acid)
Percent Purity 60.0 to 65.0% (solids)
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.23
Chemical Name or Material Poly(acrylic acid), 63 wt. % Aqueous Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.