missing translation for 'onlineSavingsMsg'
Learn More

Poly-D-Lysine Hydrobromide, MP Biomedicals

Product Code. 17318583 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17318583 50 mg 50mg
11428350 10 mg 10mg
11448350 100 mg 100mg
11458350 500 mg 500mg
11468350 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17318583 Supplier MP Biomedicals Supplier No. 0210269450
  • ONLINE EXCLUSIVE PRICE
    204.00 €
    193.20 € / 50mg
    Save 10.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 27964-99-4
Molecular Formula C6H15BrN2O2
Molecular Weight (g/mol) 227.102
InChI Key MEXAGTSTSPYCEP-NUBCRITNSA-N
Synonym d-lysine hydrobromide
PubChem CID 87493163
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrobromide
SMILES C(CCN)CC(C(=O)O)N.Br

Specifications

Quantity 50 mg
Formula Weight 4,000 to 15,000
Chemical Name or Material Poly-D-Lysine Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.