missing translation for 'onlineSavingsMsg'
Learn More

Poly-D-Lysine Hydrobromide, MP Biomedicals™

Product Code. 11466632 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466632 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11466632 Supplier MP Biomedicals Supplier No. 0215017580
  • ONLINE EXCLUSIVE PRICE
    387.00 €
    366.45 € / 100mg
    Save 20.55 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhances cell adhesion to solid substrates. Poly-D-Lysine Hydrobromide, MP Biomedicals is a synthetic amino acid that binds to DNA, red cell membrane and any negatively charged protein. It has been shown to eliminate prion proteins from infected cells. Mo

When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding. It is a compromise between easier-to-use lower molecular weight products and the extremely viscous higher molecular weights.

Poly-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. Both the D- and L- forms can be used as a coating substrate, since poly-lysine is a nonspecific attachment factor for cells; however, certain cells can digest it. In this case, poly-D-lysine should be used as the attachment factor so that the cells are not disrupted by excessive uptake of L-lysine.

Poly-D-Lysine Hydrobromide is typically used as a coating substrate for culture dishes, slides, etc. Other uses for poly-lysine have been reported as well:

  • Conjugation to methotrexate for increased drug transport
  • Microencapsulation of islets
  • Use in simple reproducible procedure for chromosomal preparations from a variety of tissues
  • Immobilation of living cells in biocompatible semipermeable microcapsules
  • In the preparation of polycationic beads
  • Conjugation to albumin and horseradish peroxidase to enhance cellular uptake

Specifications

CAS 27964-99-4
Molecular Formula C6H15BrN2O2
Synonym d-lysine hydrobromide
Solubility Information 50mg/mL, H2O
InChI Key MEXAGTSTSPYCEP-NUBCRITNSA-N
SMILES C(CCN)CC(C(=O)O)N.Br
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrobromide
Molecular Weight (g/mol) 227.102
Quantity 100 mg
PubChem CID 87493163
Formula Weight 70000-150000
Physical Form Powder
Chemical Name or Material Poly-D-lysine Hydrobromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.