missing translation for 'onlineSavingsMsg'
Learn More

Poly(dimethylsiloxane), 100 cSt

Product Code. 10143910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143910 250 g Glass bottle 250g
10521821 500 g Glass bottle 500g
10494215 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143910 Supplier Thermo Scientific Acros Supplier No. 178442500
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
Synonym Simethicone, Dimethicone, PDMS
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Physical Form Oil, Free From Suspended Matter
Linear Formula {Si(CH3)2O-}x
Density 96%5g/mL
Merck Index 10, 8374
Quantity 250 g
Infrared Spectrum Authentic
Odor Passes Test
Packaging Glass bottle
Refractive Index 1.4030 to 1.4050 (20°C, 589nm)
Viscosity 100mPa·s (25°C)
Chemical Name or Material Poly(dimethylsiloxane)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.