missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene Glycol) ~20000, TRC

Product Code. 30134733
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134733 50 g 50g
30132931 100 g 100g
17938224 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134733 Supplier Toronto Research Chemicals Supplier No. TRCP68837350G
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula H2 O(C2 H4 O)n
Molecular Weight (g/mol) 62.07
Synonym Polyethylene glycol, Polyethylene oxide, Poly(oxy-1,2-ethanediyl), α-hydro-ω-hydroxy-, Glycols, polyethylene (8CI), 1,2-Ethanediol, homopolymer, 1660O, 1660S, 400DAB8, Alcox L 11, Alkox, Alkox 8400, Alkox E 100, Alkox E 130, Alkox E 160, Alkox E 240, Alkox E 30, Alkox E 300, Alkox E 30G, Alkox E 45, Alkox E 60, Alkox E 75, Alkox E 75G, Alkox L 11, Alkox L 6, Alkox L 8, Alkox LE, Alkox R 100, Alkox R 1000, Alkox R 15, Alkox R 150, Alkox R 400, Alkox SR, Alkox SW, Antarox E 4000, Aqua Calk TWB-P, Aquacide III, Aquaffin, B 22181, BDH 301, Badimol, Beraid 3540, Bradsyn PEG, Breox 2000, Breox 20M, Breox 4000, Breox 550, Breox PEG 300, CAFO 154, CBP 20, CO 1006, Carbowax, Carbowax 100, Carbowax 1000, Carbowax 1350, Carbowax 14000, Carbowax 1450, Carbowax 1500, Carbowax 1540, Carbowax 20, Carbowax 200, Carbowax 20000, Carbowax 25000, Carbowax 300, Carbowax 3350, Carbowax 400, Carbowax 4000, Carbowax 4500, Carbowax 4600, Carbowax 600, Carbowax 6000, Carbowax 8000, Carbowax E 1000, Carbowax E 300, Carbowax E 9000, Carbowax PEG 1000, Carbowax PEG 1450, Carbowax PEG 300, Carbowax PEG 3350, Carbowax PEG 600, Carbowax PEG Plus, Carbowax Sentry 300, Carbowax Sentry NF, Carbowax Sentry PEG 8000, Cartaretin E, Cerasol 200A, Cerasol 250A, Chemiox E 20(C), Colonsoft, DB-WAX, DD 3002, Deactivator H, Decuflux RM 33, Desmophen L 1208, DuraSeal (tissue sealant), Duraseal, E 1000, E 1000 (polyglycol), E 1450NF, E 200, E 200 (polyglycol), E 240, E 30, E 300, E 3350, E 400, E 400NF, E 45, E 4500, E 4500 (polymer), E 600, E 6000, E 8000, ENT 1000, Emkapol 150, Emkapol 200, Emkapol 4200, Ethylene glycol homopolymer, Ethylene glycol polymer, Ethylene oxide polymer, Ethylene oxide, homopolymer, Ethylene polyoxide, FPR, FPR (polyether), Feltmaster 15LF, Floc 999, Flocc 999, Fomrez PEG 1000L, Forlax, G 3350, GPE 1000, GPE 400, Gafanol E 200, Gafanol E 300, GaviLyte C, Genoplast 200, Gligogum 4000, GlycoLax, H 22, H 22 (polyether), HM 500, HM 500 (polyoxyalkylene), HPEO 4250, HR 20M, IW, IW (dispersant), Isocolan, KPEG 6000, KPEG 6000LA, KleanPrep, Kollisolv PEG 300, Kollisolv PEG 400, L 300, L 6, L 6 (polyoxyalkylene), Lanogene C, Laprol 1001, Laprol 2502, Laprol 402, Lineartop P, Lipoxol 12000, Lipoxol 1500, Lipoxol 200, Lipoxol 300, Lipoxol 3000, Lipoxol 4000, Lipoxol 400MED, Lipoxol 600, Lipoxol 6000, Lixopol 600, Lumulse 1450, Lumulse PEG 200, Lutrol 300, Lutrol 400, Lutrol 75, Lutrol 9, Lutrol E 1500, Lutrol E 300, Lutrol E 400, Lutrol E 4000, Lutrol E 6000, M 2000H, M 3686B, M 9000, M-LE 1013, MN 300, Macol E 300, Macrogol, Macrogol 1000, Macrogol 1500, Macrogol 1540, Macrogol 200, Macrogol 20000, Macrogol 300, Macrogol 3000, Macrogol 400, Macrogol 4000, Macrogol 400R, Macrogol 600, Macrogol 6000, Macrogol 6000P, Macrogol 600P, Macrogol 800, Macrogol PEG 400, Maxifloc 998, Maxifloc 999, Meisei 4000, Microsolv PEG 100, MiraLax, Modopeg, Modopeg 4000, Monopol PEG 200, Muvinlax, N 80NF, Netbond FRA, Newfrontier PEM 300, Newpol PEG 600, Nongeru R 431, Nopcofloc 310, Nosilen, Nycoline, OEG, OEG 100, OEG 1000, OEG 2000, OEG 300, OEG 600, OPE 4, Optima Wax, Oxide A, Oxide Wax A, Oxide Wax AN, Oxirane polymer, Oxirane, homopolymer, Oxyethylene polymer, und weitere...
SMILES OCCO

Specifications

Quantity 50 g
Formula Weight 62.0368
Chemical Name or Material Polyethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.