missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 1450

Product Code. 10742602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
3kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742602 100 g Plastic bottle 100g
10637401 500 g Plastic bottle 500g
10187940 1 kg Plastic bottle 1kg
10557752 3 kg Plastic drum 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10742602 Supplier Thermo Scientific Acros Supplier No. 418041000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Waxy Flakes
Melting Point 44.0°C to 48.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Quantity 100 g
Hydroxyl Value 70 to 80mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Viscosity 36-42 mPa.s ( 20°C 50% in water)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol), Average M.W. 1450
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.