missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 400

Product Code. 10389123 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10389123 5 L Plastic drum 5 L
10634892 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10389123 Supplier Thermo Scientific Acros Supplier No. 192230051
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color Colorless
Physical Form Liquid
Boiling Point >200.0°C
Melting Point 4.0°C to 8.0°C
Density 1.1275g/mL
Quantity 5 L
Flash Point 235°C
Hydroxyl Value 267 to 295mg KOH/g
Packaging Plastic drum
Linear Formula H(OCH2CH2)nOH
Refractive Index 1.4650 to 1.4680
Specific Gravity 1.1275
Merck Index 15, 7688
Viscosity 110-125 mPa.s (20°C)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Formula Weight 400
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.